This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGD1305713 SIRNA Assays ›
Gene Symbol:
RGD1305713 LOC100363116
Gene Name:
similar to RIKEN cDNA 3110040N11
hypothetical protein LOC100363116
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 143601011 - 143608772 on Build Rnor_6.0
Chromosome Location:
Chr.6: 136473689 - 136475156 on Build Rnor_6.0
UniGene ID:
Rn.138288
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 252986

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1305713
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024749.1 NP_001019920.1 2 203
XM_006229429.2 XP_006229491.1 2 1908
GenBank mRNA BC094529.1 - 2 203 7320095
Gene Symbol: LOC100363116
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750235.3 XP_003750283.1 1 472

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-47mq8:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline