This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NBR1 SIRNA Assays ›
Gene Symbol:
Nbr1
Gene Name:
neighbor of Brca1 gene 1
Gene Aliases:
Ca125, RGD1311421
Chromosome Location:
Chr.10: 89455024 - 89483791 on Build Rnor_6.0
UniGene ID:
Rn.94975
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 253028

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nbr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024765.1 NP_001019936.1 12 1349
XM_006247321.2 XP_006247383.1 12 1631
XM_006247322.2 XP_006247384.1 12 1546
XM_006247323.2 XP_006247385.1 12 1403
XM_006247324.2 XP_006247386.1 12 1350
XM_006247325.2 XP_006247387.1 12 1506
XM_006247326.2 XP_006247388.1 12 1501
GenBank mRNA BC095903.1 - 12 1349 7096697
FQ211883.1 - 12 1349

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_031862, NM_031858, NM_005899, BC009808, AK308324, and others.

Gene Ontology Categories:

Function(s) Process(es)

macroautophagy
regulation of bone mineralization
regulation of stress-activated MAPK cascade
negative regulation of osteoblast differentiation
protein oligomerization
zinc ion binding
ubiquitin binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline