This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HAUS6 SIRNA Assays ›
Gene Symbol:
HAUS6
Gene Name:
HAUS augmin like complex subunit 6
Gene Aliases:
Dgt6, FAM29A
Chromosome Location:
Chr.9: 19053137 - 19102942 on Build GRCh38
UniGene ID:
Hs.533468
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 25341

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HAUS6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270890.1 NP_001257819.1 9 1334
NM_017645.4 NP_060115.3 9 1334
XM_017014830.1 XP_016870319.1 5 485
GenBank mRNA AB046794.1 - 2 190
AF533709.1 - 9 1516
AF537091.1 - 9 1296
AK023068.1 - 2 253
AK056458.1 - 9 1296
AK301852.1 - 2 194
BC026178.1 - 9 1297
BC065935.1 - 7 834
BC071625.1 - 2 293
BC111041.1 - 5 610 5529130
BC113981.1 - 9 1131 40082296
BC114492.1 - 9 1131 40082291

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
mitotic nuclear division
spindle assembly
centrosome organization
cell division
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0