siRNA Details
| Gene Symbol: ZNF280C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017666.4 | NP_060136.1 | 4, 5 | 455 | ||
| XM_006724765.3 | XP_006724828.1 | 4, 5 | 406 | |||
| GenBank mRNA | AK000102.1 | - | 4, 5 | 378 | ||
| AK290373.1 | - | 4, 5 | 406 | |||
| AL133204.1 | - | 4, 5 | 390 | |||
| AL834333.1 | - | 4, 5 | 386 | |||
| BC051728.1 | - | 4, 5 | 406 | 4828435 | ||
| DR003059.1 | - | 4, 5 | 454 | |||

ZNF280C
Genomic Map