This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TXLNGY SIRNA Assays ›
Gene Symbol:
TXLNGY
Gene Name:
taxilin gamma pseudogene, Y-linked
Gene Aliases:
CYorf15A, CYorf15B, TXLNG2P
Chromosome Location:
Chr.Y: 19567358 - 19607170 on Build GRCh38
UniGene ID:
Hs.522863
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 254497

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TXLNGY
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK090535.1 - 1 1731
AK127387.1 - 3 1444
BC110817.1 - 4 1463 6058816

Back To Top

More Information


Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline