siRNA Details
| Gene Symbol: Fbxw20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008428.3 | NP_001008428.1 | 4 | 394 | ||
| GenBank mRNA | AK135785.1 | - | 4 | 375 | ||
| AK136200.1 | - | 4 | 372 | |||
| BC085247.1 | - | 4 | 375 | 30477089 | ||
| CJ076686.1 | - | 4 | 399 | |||

Fbxw20
Genomic Map