siRNA Details
| Gene Symbol: Vom1r56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008929.1 | NP_001008929.1 | 1 | 13 | ||
| GenBank mRNA | AY510280.1 | - | 1 | 13 | ||
| Gene Symbol: LOC100909792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749304.2 | XP_003749352.1 | 1 | 66 | ||

Vom1r56
Genomic Map