This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LINC00207 SIRNA Assays ›
Gene Symbol:
LINC00207
Gene Name:
long intergenic non-protein coding RNA 207
Gene Aliases:
NCRNA00207
Chromosome Location:
Chr.22: 44569340 - 44572449 on Build GRCh38
UniGene ID:
Hs.332685
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 255243

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC00207
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028409.1 4, 5 381
NR_028410.1 3, 4 350
NR_028411.1 3, 4 314
GenBank mRNA BC144506.1 - 3, 4 338
BC144508.1 - 4, 5 369
BU585030.1 - 3, 4 333

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline