This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IL33 SIRNA Assays ›
Gene Symbol:
Il33
Gene Name:
interleukin 33
Gene Aliases:
RGD1311155
Chromosome Location:
Chr.1: 248112636 - 248147030 on Build Rnor_6.0
UniGene ID:
Rn.106849
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 255842

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Il33
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014166.1 NP_001014188.1 5 488
XM_008760344.1 XP_008758566.1 5 511
GenBank mRNA BC081993.1 - 5 488 7130307
FQ212643.1 - 5 516

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytokine production
negative regulation of leukocyte migration
negative regulation of T-helper 1 type immune response
positive regulation of type 2 immune response
transcription, DNA-templated
positive regulation of gene expression
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of interferon-gamma production
positive regulation of interleukin-13 production
positive regulation of interleukin-4 production
positive regulation of interleukin-5 production
positive regulation of interleukin-6 production
type 2 immune response
positive regulation of macrophage activation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of inflammatory response
positive regulation of immunoglobulin secretion
negative regulation of immunoglobulin secretion
defense response to virus
positive regulation of chemokine secretion
extrinsic apoptotic signaling pathway
cytokine activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline