siRNA Details
| Gene Symbol: Ndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025023.1 | NP_001020194.1 | 3 | 556 | ||
| GenBank mRNA | BC079443.1 | - | 3 | 556 | 7115485 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndc1| Gene Symbol: Ndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025023.1 | NP_001020194.1 | 3 | 556 | ||
| GenBank mRNA | BC079443.1 | - | 3 | 556 | 7115485 | |