siRNA Details
| Gene Symbol: Gys1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030678.3 | NP_109603.2 | 5 | 933 | ||
| GenBank mRNA | AK050813.1 | - | 3 | 366 | ||
| AK169344.1 | - | 5 | 929 | |||
| AK170404.1 | - | 5 | 932 | |||
| AK170585.1 | - | 5 | 853 | |||
| AK171148.1 | - | 5 | 888 | |||
| AK171426.1 | - | 5 | 912 | |||
| BC019389.1 | - | 5 | 726 | 3663506 | ||
| BC131687.1 | - | 5 | 780 | 40111722 | ||
| BC131688.1 | - | 5 | 780 | 40111723 | ||
| BC152550.1 | - | 5 | 780 | 40111724 | ||
| U53218.1 | - | 5 | 771 | |||

Gys1
Genomic Map