This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RAB20 SIRNA Assays ›
Gene Symbol:
Rab20
Gene Name:
RAB20, member RAS oncogene family
Gene Aliases:
AA536966, D8Ertd350e
Chromosome Location:
Chr.8: 11453513 - 11478499 on Build GRCm38
UniGene ID:
Mm.37632
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 257439

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011227.1 NP_035357.1 1 47
GenBank mRNA AB232614.1 - 1 47
AK145566.1 - 1 224
AK152454.1 - 1 173
AK163743.1 - 1 187
AK164541.1 - 1 187
BC051569.1 - 1 265
BC089570.1 - 1 172 6747570
M79310.1 - 1 47
X80332.1 - 1 47

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109535.

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
small GTPase mediated signal transduction
protein transport
regulation of endocytosis
Rab protein signal transduction
phagosome acidification
phagosome-lysosome fusion
nucleotide binding
GTPase activity
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline