siRNA Details
| Gene Symbol: 1600002K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027207.2 | NP_081483.1 | 3 | 413 | ||
| GenBank mRNA | AK005397.1 | - | 3 | 414 | ||
| AK050606.1 | - | 3 | 412 | |||
| AK168176.1 | - | 3 | 411 | |||
| BC146018.1 | - | 3 | 411 | 40131321 | ||
| BC146020.1 | - | 3 | 411 | 40131323 | ||

1600002K03Rik
Genomic Map