This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DCAF11 SIRNA Assays ›
Gene Symbol:
Dcaf11
Gene Name:
DDB1 and CUL4 associated factor 11
Gene Aliases:
Wdr23
Chromosome Location:
Chr.15: 34233851 - 34244512 on Build Rnor_6.0
UniGene ID:
Rn.16542
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 259126

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dcaf11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009686.2 NP_001009686.1 15 2281
XM_006251971.2 XP_006252033.1 16 2616
GenBank mRNA BC081957.1 - 16 2486
BC087110.1 - 14 2062 7193336
BC091364.1 - 15 2281 7319156
FQ222851.1 - 16 2582

Back To Top

More Information


Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s)

protein ubiquitination

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-bvzv4:80/100.66.75.57:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0