siRNA Details
| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013133.1 | NP_001013151.1 | 4 | 776 | ||
| GenBank mRNA | BC091309.1 | - | 4 | 776 | 7311705 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhobtb2| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013133.1 | NP_001013151.1 | 4 | 776 | ||
| GenBank mRNA | BC091309.1 | - | 4 | 776 | 7311705 | |