This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RMDN3 SIRNA Assays ›
Gene Symbol:
Rmdn3 LOC100911313
Gene Name:
regulator of microtubule dynamics 3
regulator of microtubule dynamics protein 3-like
Gene Aliases:
Fam82a2, Fam82c, Ptpip51, RGD1308697, Rmd-3
-
Chromosome Location:
Chr.3: 110943843 - 110964449 on Build Rnor_6.0
Chromosome Location:
Chr.3: 112605090 - 112616301 on Build Rnor_6.0
UniGene ID:
Rn.18772
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 260293

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rmdn3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014046.2 NP_001014068.1 12 1410
GenBank mRNA BC082081.1 - 12 1372 7133344
Gene Symbol: LOC100911313
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762136.1 XP_008760358.1 9 1090

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cellular calcium ion homeostasis
apoptotic process
cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-qjpw4:80/100.66.76.224:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0