siRNA Details
| Gene Symbol: Luc7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024269.1 | NP_001019440.1 | 4, 5 | 684 | ||
| XM_006246006.2 | XP_006246068.1 | 5, 6 | 649 | |||
| XM_006246007.1 | XP_006246069.1 | 5, 6 | 2053 | |||
| XM_008767588.1 | XP_008765810.1 | 5, 6 | 1512 | |||
| GenBank mRNA | BC089960.1 | - | 4, 5 | 684 | 7313914 | |

Luc7l
Genomic Map