This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGD1310852 SIRNA Assays ›
Gene Symbol:
RGD1310852
Gene Name:
similar to RIKEN cDNA 9130401M01
Gene Aliases:
-
Chromosome Location:
Chr.7: 97891723 - 97902585 on Build Rnor_6.0
UniGene ID:
Rn.159189
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 260936

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1310852
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025007.1 NP_001020178.1 5 914
GenBank mRNA BC091232.1 - 5 914 7322456

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029418, AK089715, AK018655, BC050895, AK047927, and others.

Panther Classification:

Molecular Function -

homeodomain transcription factor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline