siRNA Details
| Gene Symbol: RAB11FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002814.2 | NP_001002814.2 | 6 | 4298 | ||
| NM_025151.4 | NP_079427.4 | 5 | 2396 | |||
| GenBank mRNA | AK092296.1 | - | 4 | 1704 | ||
| AK122583.1 | - | 3 | 2437 | |||
| AK124422.1 | - | 1 | 1196 | |||
| AK226111.1 | - | 6 | 3988 | |||
| AK291781.1 | - | 5 | 2393 | |||
| AL832155.1 | - | 3 | 1879 | |||
| AL833169.1 | - | 3 | 1708 | |||
| BC077720.1 | - | 5 | 2366 | 6537110 | ||

RAB11FIP1
Genomic Map