This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 9530068E07RIK SIRNA Assays ›
Gene Symbol:
9530068E07Rik
Gene Name:
RIKEN cDNA 9530068E07 gene
Gene Aliases:
C85061, Kct2
Chromosome Location:
Chr.11: 52396428 - 52408723 on Build GRCm38
UniGene ID:
Mm.291979
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 261574

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 9530068E07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153117.2 NP_694757.1 3 1915
GenBank mRNA AK079269.1 - 3 1916
AK144677.1 - 3 1881
AK145254.1 - 3 1910
AK147956.1 - 3 1880
AK150517.1 - 3 1899
AK154955.1 - 3 1885
BC006957.1 - 3 1769
BC034829.1 - 3 1889 4482756

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fh6lz:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline