This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 9530068E07RIK SIRNA Assays ›
Gene Symbol:
9530068E07Rik
Gene Name:
RIKEN cDNA 9530068E07 gene
Gene Aliases:
C85061, Kct2
Chromosome Location:
Chr.11: 52396428 - 52408723 on Build GRCm38
UniGene ID:
Mm.291979
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 261575

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 9530068E07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153117.2 NP_694757.1 3 1558
GenBank mRNA AK079269.1 - 3 1559
AK144677.1 - 3 1523
AK147956.1 - 3 1525
AK150517.1 - 3 1542
AK154955.1 - 3 1528
BC006957.1 - 3 1412
BC034829.1 - 3 1532 4482756

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline