siRNA Details
| Gene Symbol: Itgbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017505.1 | NP_001017505.1 | 11 | 1709 | ||
| GenBank mRNA | BC087076.1 | - | 11 | 1709 | 7134947 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgbl1| Gene Symbol: Itgbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017505.1 | NP_001017505.1 | 11 | 1709 | ||
| GenBank mRNA | BC087076.1 | - | 11 | 1709 | 7134947 | |