This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MRC2 SIRNA Assays ›
Gene Symbol:
Mrc2
Gene Name:
mannose receptor, C type 2
Gene Aliases:
Endo180
Chromosome Location:
Chr.10: 93520311 - 93581599 on Build Rnor_6.0
UniGene ID:
Rn.213511
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 261982

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mrc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024687.1 NP_001019858.1 21 3002
XM_006247631.2 XP_006247693.1 21 3127
GenBank mRNA DQ058624.1 - 21 3002

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006039, BC153884, AK093230, BC150212, BC146647, and others.
This siRNA also targets the following transcripts in Mouse: NM_008626, AK129195, BC116642, BU053305, U56734.

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
endocytosis
collagen catabolic process
collagen binding
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-6l2j2:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline