siRNA Details
| Gene Symbol: Ccdc71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024904.1 | NP_001020075.1 | 2 | 360 | ||
| XM_006243832.1 | XP_006243894.1 | 2 | 389 | |||
| XM_006243835.2 | XP_006243897.1 | 2 | 429 | |||
| GenBank mRNA | BC098020.1 | - | 2 | 360 | 7455232 | |

Ccdc71
Genomic Map