siRNA Details
| Gene Symbol: Fam71b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025031.1 | NP_001020202.1 | 2 | 1157 | ||
| GenBank mRNA | BC082044.1 | - | 2 | 1157 | 7131685 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam71b| Gene Symbol: Fam71b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025031.1 | NP_001020202.1 | 2 | 1157 | ||
| GenBank mRNA | BC082044.1 | - | 2 | 1157 | 7131685 | |