This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZPBP SIRNA Assays ›
Gene Symbol:
Zpbp
Gene Name:
zona pellucida binding protein
Gene Aliases:
-
Chromosome Location:
Chr.14: 91359751 - 91556306 on Build Rnor_6.0
UniGene ID:
Rn.144060
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 262029

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zpbp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025139.1 NP_001020310.1 4 625
XM_006251505.2 XP_006251567.1 4 655
GenBank mRNA BC079315.1 - 4 625 7134936

Back To Top

More Information


Gene Ontology Categories:

Function(s)

acrosome assembly
binding of sperm to zona pellucida

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline