siRNA Details
| Gene Symbol: Prl7b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029355.2 | NP_083631.1 | 2, 3 | 264 | ||
| GenBank mRNA | AF525156.1 | - | 2, 3 | 264 | ||
| AK005458.1 | - | 2, 3 | 265 | |||
| BC116772.1 | - | 2, 3 | 241 | 40126091 | ||
| BC119183.1 | - | 2, 3 | 241 | 8733932 | ||

Prl7b1
Genomic Map