This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other INTS10 SIRNA Assays ›
Gene Symbol:
INTS10
Gene Name:
integrator complex subunit 10
Gene Aliases:
C8orf35, INT10
Chromosome Location:
Chr.8: 19817149 - 19852078 on Build GRCh38
UniGene ID:
Hs.512627
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 26297

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: INTS10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018142.2 NP_060612.2 7, 8 968
XM_005273555.3 XP_005273612.1 7, 8 1226
XM_005273556.3 XP_005273613.1 7, 8 1010
XM_005273557.3 XP_005273614.1 7, 8 1010
XM_006716355.2 XP_006716418.1 7, 8 1115
XM_017013606.1 XP_016869095.1 7, 8 1010
XM_017013607.1 XP_016869096.1 7, 8 1226
GenBank mRNA AK001431.1 - 7, 8 888
BC001915.1 - 5, 6 605
BC006209.2 - 5, 6 605
BC051282.1 - 5, 6 619
CR457262.1 - 4, 5 498

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline