This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RMDN3 SIRNA Assays ›
Gene Symbol:
RMDN3
Gene Name:
regulator of microtubule dynamics 3
Gene Aliases:
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
Chromosome Location:
Chr.15: 40735884 - 40755336 on Build GRCh38
UniGene ID:
Hs.511067
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 26300

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RMDN3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304802.1 NP_001291731.1 4 661
NM_001323894.1 NP_001310823.1 4 575
NM_001323895.1 NP_001310824.1 4 792
NM_001323896.1 NP_001310825.1 4 575
NM_001323897.1 NP_001310826.1 4 691
NM_018145.2 NP_060615.1 4 691
GenBank mRNA AB000782.1 - 3 430
AK001441.1 - 4 615
AK092058.1 - 4 574
AK097286.1 - 5 503
AK123192.1 - 5 569
AK123282.1 - 3 1498
AY358793.1 - 3 431
BC063844.1 - 4 579 6527521
DA280017.1 - 4 547

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
apoptotic process
cell differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-nrgn4:80/100.66.79.50:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline