siRNA Details
| Gene Symbol: A730008H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172505.4 | NP_766093.1 | 3, 4 | 301 | ||
| GenBank mRNA | AK042589.1 | - | 3, 4 | 301 | ||
| BC131919.1 | - | 3, 4 | 269 | 40130196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A730008H23Rik| Gene Symbol: A730008H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172505.4 | NP_766093.1 | 3, 4 | 301 | ||
| GenBank mRNA | AK042589.1 | - | 3, 4 | 301 | ||
| BC131919.1 | - | 3, 4 | 269 | 40130196 | ||