siRNA Details
| Gene Symbol: LINC00310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027267.1 | 2 | 237 | |||
| GenBank mRNA | AF426258.1 | - | 2 | 237 | ||
| BC093749.1 | - | 2 | 167 | |||
| BC111996.1 | - | 2 | 167 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00310| Gene Symbol: LINC00310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027267.1 | 2 | 237 | |||
| GenBank mRNA | AF426258.1 | - | 2 | 237 | ||
| BC093749.1 | - | 2 | 167 | |||
| BC111996.1 | - | 2 | 167 | |||