This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ADGB SIRNA Assays ›
Gene Symbol:
ADGB
Gene Name:
androglobin
Gene Aliases:
C6orf103, CAPN16
Chromosome Location:
Chr.6: 146598965 - 146815462 on Build GRCh38
UniGene ID:
Hs.720870
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 263536

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ADGB
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024694.3 NP_078970.3 6 731
XM_006715566.3 XP_006715629.1 6 766
XM_011536125.2 XP_011534427.1 6 717
XM_011536126.1 XP_011534428.1 6 766
XM_011536127.2 XP_011534429.1 6 766
XM_011536128.2 XP_011534430.1 6 766
XM_011536130.2 XP_011534432.1 7 665
XM_017011314.1 XP_016866803.1 6 766
XM_017011315.1 XP_016866804.1 6 766
GenBank mRNA AK097570.1 - 6 731

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
calcium-dependent cysteine-type endopeptidase activity
oxygen binding
heme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0