This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BMP8A SIRNA Assays ›
Gene Symbol:
BMP8A
Gene Name:
bone morphogenetic protein 8a
Gene Aliases:
-
Chromosome Location:
Chr.1: 39491626 - 39529869 on Build GRCh38
UniGene ID:
Hs.472497
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 263558

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BMP8A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181809.3 NP_861525.2 7 5363
GenBank mRNA AK024413.1 - 1 1781
AK127199.1 - 2 2290

Back To Top

More Information


Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

ossification
diet induced thermogenesis
positive regulation of pathway-restricted SMAD protein phosphorylation
BMP signaling pathway
growth
regulation of apoptotic process
regulation of MAPK cascade
negative regulation of insulin secretion
cell development
cartilage development
SMAD protein signal transduction
regulation of energy homeostasis
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
BMP receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-qdm6g:80/100.66.79.8:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline