This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SMU1 SIRNA Assays ›
Gene Symbol:
SMU1
Gene Name:
DNA replication regulator and spliceosomal factor
Gene Aliases:
BWD, SMU-1, fSAP57
Chromosome Location:
Chr.9: 33041852 - 33076716 on Build GRCh38
UniGene ID:
Hs.655351
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 26360

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SMU1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018225.2 NP_060695.2 2 209
XM_005251503.4 XP_005251560.1 2 198
GenBank mRNA AK001667.1 - 2 139
AK225559.1 - 2 164
AK225587.1 - 2 158
BC002876.2 - 2 155 3940669
BC110654.1 - 2 249 6168993
EF011613.1 - 2 155

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-8dlb9:80/100.66.79.146:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline