siRNA Details
| Gene Symbol: MUC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164462.1 | NP_001157934.1 | 2 | 14944 | ||
| GenBank mRNA | AF147790.1 | - | 1 | 696 | ||
| AK024856.1 | - | 1 | 451 | |||
| BC127877.1 | - | 1 | 714 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MUC12| Gene Symbol: MUC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164462.1 | NP_001157934.1 | 2 | 14944 | ||
| GenBank mRNA | AF147790.1 | - | 1 | 696 | ||
| AK024856.1 | - | 1 | 451 | |||
| BC127877.1 | - | 1 | 714 | |||