siRNA Details
| Gene Symbol: ISM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080826.1 | NP_543016.1 | 6 | 1295 | ||
| GenBank mRNA | AK307409.1 | - | 4 | 734 | ||
| BC017997.1 | - | 1 | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ISM1| Gene Symbol: ISM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080826.1 | NP_543016.1 | 6 | 1295 | ||
| GenBank mRNA | AK307409.1 | - | 4 | 734 | ||
| BC017997.1 | - | 1 | 351 | |||