siRNA Details
| Gene Symbol: LOC145845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024264.1 | 7 | 1945 | |||
| GenBank mRNA | AK023254.1 | - | 5 | 1645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC145845| Gene Symbol: LOC145845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024264.1 | 7 | 1945 | |||
| GenBank mRNA | AK023254.1 | - | 5 | 1645 | ||