siRNA Details
Gene Symbol: RGS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039152.3 | NP_001034241.1 | 3 | 253 | ||
GenBank mRNA | AY643711.1 | - | 3 | 254 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039152.3 | NP_001034241.1 | 3 | 253 | ||
GenBank mRNA | AY643711.1 | - | 3 | 254 |