siRNA Details
| Gene Symbol: SMU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018225.2 | NP_060695.2 | 2 | 299 | ||
| XM_005251503.4 | XP_005251560.1 | 2 | 288 | |||
| GenBank mRNA | AK001667.1 | - | 2 | 229 | ||
| AK225559.1 | - | 2 | 254 | |||
| AK225587.1 | - | 2 | 248 | |||
| BC002876.2 | - | 2 | 245 | 3940669 | ||
| EF011613.1 | - | 2 | 245 | |||

SMU1
Genomic Map