siRNA Details
| Gene Symbol: MUC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164462.1 | NP_001157934.1 | 2 | 5225 | ||
| GenBank mRNA | AF147791.1 | - | 1 | 479 | ||
| AK025404.1 | - | 1 | 548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MUC12| Gene Symbol: MUC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164462.1 | NP_001157934.1 | 2 | 5225 | ||
| GenBank mRNA | AF147791.1 | - | 1 | 479 | ||
| AK025404.1 | - | 1 | 548 | |||