This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SMCO2 SIRNA Assays ›
Gene Symbol:
SMCO2
Gene Name:
single-pass membrane protein with coiled-coil domains 2
Gene Aliases:
C12orf70
Chromosome Location:
Chr.12: 27446482 - 27506911 on Build GRCh38
UniGene ID:
Hs.282121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 265794

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SMCO2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145010.1 NP_001138482.1 8 877
XM_011520634.2 XP_011518936.1 8 1055
XM_011520635.2 XP_011518937.1 6 554
XM_011520636.2 XP_011518938.1 6 910
XM_017019254.1 XP_016874743.1 6 811
XM_017019255.1 XP_016874744.1 5 661

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0