siRNA Details
| Gene Symbol: LINC00374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132367.1 | 5, 6 | 619 | |||
| GenBank mRNA | BG190276.1 | - | 5, 6 | 589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00374| Gene Symbol: LINC00374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132367.1 | 5, 6 | 619 | |||
| GenBank mRNA | BG190276.1 | - | 5, 6 | 589 | ||