This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF444 SIRNA Assays ›
Gene Symbol:
ZNF444
Gene Name:
zinc finger protein 444
Gene Aliases:
EZF-2, EZF2, ZSCAN17
Chromosome Location:
Chr.19: 56140951 - 56160893 on Build GRCh38
UniGene ID:
Hs.24545
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 26638

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF444
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253792.1 NP_001240721.1 4 777
NM_018337.3 NP_060807.2 4 777
XM_005259034.2 XP_005259091.1 5 882
XM_005259035.1 XP_005259092.1 4 814
XM_005259036.1 XP_005259093.1 4 681
XM_011527068.1 XP_011525370.1 4 840
XM_011527069.1 XP_011525371.1 5 732
XM_011527070.1 XP_011525372.1 5 2426
XM_011527071.1 XP_011525373.1 5 853
XM_011527073.1 XP_011525375.1 5 2426
XM_017026932.1 XP_016882421.1 5 715
XM_017026933.1 XP_016882422.1 2 1026
GenBank mRNA AB052954.1 - 4 620
AB052955.1 - 4 620
AK001999.1 - 4 622
AK074063.1 - 1 4332
AK097319.1 - 1 2005
AK223408.1 - 4 647
AL834130.1 - 4 757
BC021282.2 - 4 607 5016494

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0