siRNA Details
| Gene Symbol: SPDYE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128076.1 | NP_001121548.1 | 1 | 260 | ||
| XM_005256632.4 | XP_005256689.1 | 1 | 1129 | |||
| XM_005256633.4 | XP_005256690.1 | 1 | 1129 | |||
| GenBank mRNA | BC146949.1 | - | 1 | 260 | 9056877 | |
| BC146966.1 | - | 1 | 260 | 9056894 | ||

SPDYE4
Genomic Map