This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF654 SIRNA Assays ›
Gene Symbol:
ZNF654
Gene Name:
zinc finger protein 654
Gene Aliases:
-
Chromosome Location:
Chr.3: 88059217 - 88144664 on Build GRCh38
UniGene ID:
Hs.591650
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 26697

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF654
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018293.2 NP_060763.2 1 399
XM_011534340.2 XP_011532642.2 7 1803
XM_017006789.1 XP_016862278.1 8 1932
XM_017006790.1 XP_016862279.1 8 1944
XM_017006791.1 XP_016862280.1 6 1583
XM_017006792.1 XP_016862281.1 6 1584
XM_017006793.1 XP_016862282.1 7 1803
XM_017006794.1 XP_016862283.1 6 1543
XM_017006795.1 XP_016862284.1 7 1737
GenBank mRNA AF543494.1 - 1 400
AK001859.1 - 1 186

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0