siRNA Details
| Gene Symbol: LINC00578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047568.1 | 4 | 705 | |||
| GenBank mRNA | DB235147.1 | - | 2 | 424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00578| Gene Symbol: LINC00578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047568.1 | 4 | 705 | |||
| GenBank mRNA | DB235147.1 | - | 2 | 424 | ||