This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HEMGN SIRNA Assays ›
Gene Symbol:
HEMGN
Gene Name:
hemogen
Gene Aliases:
CT155, EDAG, EDAG-1, NDR
Chromosome Location:
Chr.9: 97926791 - 97944915 on Build GRCh38
UniGene ID:
Hs.176626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 26899

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HEMGN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018437.4 NP_060907.2 4 974
NM_197978.2 NP_932095.1 3 1028
XM_005252086.1 XP_005252143.1 4 936
XM_011518845.2 XP_011517147.1 4 848
XM_011518846.2 XP_011517148.1 4 958
GenBank mRNA AF116617.1 - 1 430
AF130060.2 - 3 761
AF228713.2 - 4 881
AF306682.1 - 3 810
AF306683.1 - 3 776
AF322875.1 - 4 880
AK292441.1 - 4 900
AL833094.1 - 3 820
AY244805.1 - 3 974
AY255672.1 - 3 879
BC048324.1 - 4 883 5166612

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
cell differentiation
regulation of osteoblast differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0