This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HEMGN SIRNA Assays ›
Gene Symbol:
HEMGN
Gene Name:
hemogen
Gene Aliases:
CT155, EDAG, EDAG-1, NDR
Chromosome Location:
Chr.9: 97926791 - 97944915 on Build GRCh38
UniGene ID:
Hs.176626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 26994

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HEMGN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018437.4 NP_060907.2 4 1057
NM_197978.2 NP_932095.1 3 1111
XM_005252086.1 XP_005252143.1 4 1019
XM_011518845.2 XP_011517147.1 4 931
XM_011518846.2 XP_011517148.1 4 1041
GenBank mRNA AF116617.1 - 1 513
AF130060.2 - 3 844
AF228713.2 - 4 964
AF306682.1 - 3 893
AF306683.1 - 3 859
AF322875.1 - 4 963
AK292441.1 - 4 983
AL833094.1 - 3 903
AY244805.1 - 3 1057
AY255672.1 - 3 962
BC048324.1 - 4 966 5166612

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
cell differentiation
regulation of osteoblast differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline