This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MEIKIN SIRNA Assays ›
Gene Symbol:
MEIKIN
Gene Name:
meiotic kinetochore factor
Gene Aliases:
-
Chromosome Location:
Chr.5: 131806990 - 131945698 on Build GRCh38
UniGene ID:
Hs.631802
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 270760

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MEIKIN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303622.1 NP_001290551.1 6 748
GenBank mRNA AB987829.1 - 6 555
DY654845.1 - 2 147
DY655061.1 - 5 433

Back To Top

More Information


Gene Ontology Categories:

Function(s)

male meiosis chromosome segregation
meiotic sister chromatid cohesion involved in meiosis I
female meiosis chromosome segregation
homologous chromosome segregation
meiotic sister chromatid cohesion, centromeric

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline