This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RNF130 SIRNA Assays ›
Gene Symbol:
RNF130
Gene Name:
ring finger protein 130
Gene Aliases:
G1RZFP, GOLIATH, GP
Chromosome Location:
Chr.5: 179911651 - 180072130 on Build GRCh38
UniGene ID:
Hs.484363
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 27085

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF130
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018434.5 NP_060904.2 8, 9 1669
GenBank mRNA AF155650.1 - 7, 8 968
AY083998.1 - 8, 9 1651
BC017100.1 - 8, 9 1251
BC065244.1 - 7, 8 914
BC082267.1 - 8, 9 1266
BC108306.1 - 8, 9 1664 6137501
BC113864.1 - 9, 10 1099 40033418
BC114472.1 - 8, 9 1031

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001037658, AY190520, BC166595, FQ234228.
This siRNA also targets the following transcripts in Mouse: NM_021540, AF171875, AK191139, BC018199, BC048901.

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
programmed cell death
protein ubiquitination
ubiquitin-protein transferase activity
zinc ion binding
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline